HEPTADECANOIC ACID-METHYL ESTER 0.02% 1 * 10 items
CMPLRD091158L1.10
Frequently Bought Together
pH indicator papers, rolls, Whatman™
: Whatman products (Cytiva)
Formaldehyde 37% (w/w) in water stabilised
: Thermo Fisher Scientific
Lugol's iodine solution stabilised
: VWR Chemicals
Sodium hydroxide, anhydrous ≥98%, pellets, Reagent Grade
: Honeywell Chemicals