Order Entry
Italy
Orders LinkContactUsLinkComponent
Hei-PLATE Sensor Packages Expert
Hei-PLATE Sensor Packages Expert
Hei-PLATE sensor advanced package expert
Catalog # HEID506-31151-00
Hei-PLATE Sensor Packages Expert
Catalog # HEID506-31151-00
Hei-PLATE Sensor Packages Expert
Hei-PLATE Sensor Packages Expert
Hei-PLATE sensor advanced package expert
Price
1 740,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Hei-PLATE sensor Basic and Advanced package Expert.
Basic package includes: Magnetic stirrer Hei-PLATE Mix'n'Heat Expert (145 mm diameter) - Fast and precise heating - Without overshoot - for sample-friendly work, and Pt1000 temperature sensor for precise temperature regulation.
Advanced package includes: Magnetic stirrer Hei-PLATE Mix'n'Heat Expert (145 mm diameter) - Fast and precise heating - Without overshoot - for sample-friendly work, Pt1000 temperature sensor for precise temperature regulation, Pt1000 holder with cable device for securely attaching the temperature sensor and silicone protective cover for a longer lasting protection of your magnetic stirrer.
Specifications
  • Description:
    Hei-PLATE sensor advanced package expert
  • Seq:
    02