Order Entry
Italy
ContactUsLinkComponent
Fully automatic Laboratory autoclave. Chamber Dimensions: Diameter 38cm Depth 69cm External Dimensions: 72cm (W) x 137cm (H) x 119cm (D). Chamber Volume: 85L Made of St. St. 316L 1 * 1 items
Catalog # TUTT3870ELCPVGP380
Supplier:  TUTTNAUER
Fully automatic Laboratory autoclave. Chamber Dimensions: Diameter 38cm Depth 69cm External Dimensions: 72cm (W) x 137cm (H) x 119cm (D). Chamber Volume: 85L Made of St. St. 316L 1 * 1 items
Catalog # TUTT3870ELCPVGP380
Supplier:  TUTTNAUER
Frequently Bought Together