Objective LD A-Plan 40x/0.55 Ph2 M27 (FWD=2.3mm at CG=1mm polystyrene) 1 * 1 items
Catalog # ZEIS4212619918000
Supplier: ZEISS
Objective LD A-Plan 40x/0.55 Ph2 M27 (FWD=2.3mm at CG=1mm polystyrene) 1 * 1 items
Catalog # ZEIS4212619918000
Supplier: ZEISS
Frequently Bought Together
Accessories for VWR® V-1200, UV-1600PC, UV-3100PC
Supplier: COLLECTION PREFIX HPLC
VWR® PH 20 pH Meter, Handheld
Supplier: VWR Collection
Accessories for titrators
Supplier: Mettler - Toledo
Conductivity standards
Supplier: WTW
pH/mV/°C meters, bench, inoLab® pH 7110 / 7310
Supplier: WTW
VWR® pHenomenal® Electrode Stand
Supplier: VWR Collection
VWR® pHenomenal® pH 1100 H pH/mV/°C Meter, Handheld
Supplier: VWR Collection
Titration beakers
Supplier: Mettler - Toledo
Accessories for Mettler balances
Supplier: Mettler - Toledo
VWR® HCO 304, Conductivity Meter, Handheld
Supplier: VWR Collection
Gas detection tubes for water vapour
Supplier: Dräger
Accessories for OxiTop® and OxiTop® Control
Supplier: WTW
Accessories for cell counter, EVE™
Supplier: NANOENTEK
VWR®, Hand Tally Counter
Supplier: VWR Collection
Combined titration electrodes, plug and play
Supplier: Mettler - Toledo
Flow Cells for MinION™ and GridION™ Devices
Supplier: Oxford Nanopore Technologies
Plastic cuvette for LICO
Supplier: Hach