GPC STANDARD DEXTRAN 150 1 * 800 mg
Catalog # MSPP552201500001
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffer solutions, NIST/DIN
Supplier: Mettler - Toledo
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
Pesticide reference standard, Chlorpyrifos 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Supplier: EHRENSTORFER
100 µg/mL in Acetonitrile:Dimethyl sulfoxide 1 * 1 mL
Supplier: EHRENSTORFER
Supelco® TraceCERT® Organic Reference Standard, Tebutam
Supplier: Merck
Azadirachtin certified 1 * 5 mg
Supplier: EHRENSTORFER
Lithium standard for IC, TraceCERT®, Supelco®
Supplier: Merck
Pesticide reference standard, 4-Chloroaniline 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Supplier: EHRENSTORFER
Supelco® Organic Reference Standard, Acrylonitrile
Supplier: Merck
Diflufenican certified 1 * 0,1 g
Supplier: EHRENSTORFER
4-ISOPROPYLPHENOL 1 * 50 mg
Supplier: Merck
Accessories for ion selective meters, LAQUAtwin
Supplier: HORIBA
pH Buffer Solution pH 7.00 ± 0.02 ,25°C (Coloured) 1 * 500 mL
Supplier: REAGECON
Buffer solution pH 5, Acetate Buffer
Supplier: Thermo Fisher Scientific