GPC STANDARD DEXTRAN 5 1 * 800 mg
Catalog # MSPP552200050001
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Accessories for pH/°C meter, handheld
Supplier: TESTO
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR®, Sucrose (Brix) standards, stabilised solutions for refractometry
Supplier: VWR Chemicals
VWR®, Sulphate 1000 mg/L in water standard for ion chromatography
Supplier: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals