0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Italy
ContactUsLinkComponent
10G PL COPPER 1 (71 TESTS) 1 * 10 g
WTIDPLPOW10CU1
:
WATER ID PRIMELAB
10G PL COPPER 1 (71 TESTS) 1 * 10 g
WTIDPLPOW10CU1
:
WATER ID PRIMELAB
Frequently Bought Together
REAGENT KIT FOR PRIMELAB ID71(430 TESTS) 1 * 1 items
: WATER ID PRIMELAB
MES MONOHYDRATE CGMP MANUFACTURED 1 * 12 kg
: Spectrum Chemical
Sodium hydroxide, anhydrous ≥98%, pellets, Reagent Grade
: Honeywell Chemicals
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
: Hach