0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Italy
ContactUsLinkComponent
65ML PL OXYGEN SCAVENGER 1 (430 TESTS) 1 * 65 mL
WTIDPL65OXYSCAV1
:
WATER ID PRIMELAB
65ML PL OXYGEN SCAVENGER 1 (430 TESTS) 1 * 65 mL
WTIDPL65OXYSCAV1
:
WATER ID PRIMELAB
Frequently Bought Together
EOSIN RAL 555 1L 1 * 1 L
: RAL
Test kits, chlorine, Spectroquant®, Supelco®
: Merck
Fluorescein diacetate cell viability stain, Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
Kjeldahl Catalyst (Cu-Se) (1.5% CuSO4.5H2O + 2% Se) tablets 1 * 5 kg
: PanReac AppliChem
Ferrozine mono-sodium salt hydrate
: Hach
Reagents for DR800, DR900, DR2010, DR2500, DR2400, DR2800, DR3800, DR3900, DR5000, DR6000
: Hach
Anti-IL-10 Rabbit Recombinant Multiclonal Antibody [clone: RP23040032]
: Abcam