Volumetric flask, PMP, transparent, 50 ml, NS 12/21, PP-stopper 1 * 2 items
Catalog # BRND36174
Supplier: Brand
Volumetric flask, PMP, transparent, 50 ml, NS 12/21, PP-stopper 1 * 2 items
Catalog # BRND36174
Supplier: Brand
Frequently Bought Together
Benzyl alcohol ≥99.0% analytical reagent
Supplier: VWR Chemicals
VWR®, Bottles, Narrow Neck, Round, LDPE, with Screw Caps
Supplier: VWR Collection
Sulphuric acid 95,0 - 97,0% 95.0-97.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Pipette tips
Supplier: Sartorius
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Pipette tips, epT.I.P.S.® Standard (General Lab Product)
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
NOZZLE ASSY-ASXL FID 1 * 1 items
Supplier: PerkinElmer