0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Italy
ContactUsLinkComponent
CONDUCTIVITY PROBE WITH ATC, GLASS K=1 (AS SUPPLIED WITH 4510, 4520 AND 3540) 1 * 1 items
JENW99968-36
:
Cole-Parmer
CONDUCTIVITY PROBE WITH ATC, GLASS K=1 (AS SUPPLIED WITH 4510, 4520 AND 3540) 1 * 1 items
JENW99968-36
:
Cole-Parmer
Frequently Bought Together
pH electrodes, InLab® ISM
: Mettler - Toledo
pH micro electrodes, InLab® Micro Pro ISM® / InLab® Ultra-Micro-ISM® / InLab® Surface Pro-ISM
: Mettler - Toledo
Wipes for general applications, Tork Premium, colour coded
: Essity
Ion selective electrode, Ammonia, solid-state, IntelliCAL™
: Hach
WIPER POLYNIT HEATSEAL 5X5CM BULK 1 * 12.500 items
: CONTEC
Cleanroom wipes, TechniCloth®
: Texwipe
Ion selective electrodes, chloride, Orion™
: Thermo Orion
Cleanroom wipes, sterile, Polynit Heatseal
: CONTEC
Quick spins, DBI-SALA®, 3M™
: CAPITAL SAFETY
Combined ion-selective electrodes, perfectION™
: Mettler - Toledo
Cables
: Mettler - Toledo