Geranyl acetate
Geranyl acetate
Catalog # EHERC14010500
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC(=CCC\C(=C\COC(=O)C)\C)C
- Inchi: InChI=1S/C12H20O2/c1-10(2)6-5-7-11(3)8-9-14-12(4)13/h6,8H,5,7,9H2,1-4H3/b11-8+
- IUPAC: [(2E)-3,7-dimethylocta-2,6-dienyl] acetate
Specifications
Specifications
- Description:Geranyl acetate
- Cat. No.:EHERC14010500
- Formula:C₁₂H₂₀O₂
- Storage conditions:–18 °C
- Hazard class description:GHS07, GHS09
- GHS signal:Warning
- Synonym(s):2,6-Octadien-1-ol, 3,7-dimethyl-, 1-acetate, (2E)-; 2,6-Octadien-1-ol, 3,7-dimethyl-, acetate, (2E)- (9CI); 2,6-Octadien-1-ol, 3,7-dimethyl-, acetate, (E)- (8CI); Geraniol acetate (6CI); (E)-3,7-Dimethyl-2,6-octadien-1-ol acetate; (E)-3,7-Dimethyl-2,6-octadienyl acetate; 3,7-Dimethyl-2,6-octadien-1-ol acetate; Acetic acid (2E)-3,7-dimethyl-2,6-octadienyl ester; Acetic acid geraniol ester; Bay pine (oyster) oil; Geranyl acetate; Geranyl ethanoate; NSC 2584; trans-1-Acetoxy-3,7-dimethyl-2,6-octadiene; trans-3,7-Dimethyl-2,6-octadien-1-yl acetate; trans-Geranyl acetate; β-Geranyl acetate
- Molecular weight:196,29
- CAS No.:105-87-3
- Shipping conditions:20 °C
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



