Order Entry
Italy
ContactUsLinkComponent
SHODEX GPC HK-406, ULTRA-RAPID SIZE EXCLUSION COLUMN, 4,6x150mm, 6,5µm, 10,000Å, THF 1 * 1 items
Catalog # SHOWF6025060
Supplier:  Resonac
SHODEX GPC HK-406, ULTRA-RAPID SIZE EXCLUSION COLUMN, 4,6x150mm, 6,5µm, 10,000Å, THF 1 * 1 items
Catalog # SHOWF6025060
Supplier:  Resonac
Frequently Bought Together