TRITON X100 REPLACEMENT TERGITOL 15S9 1 * 500 mL
Catalog # APPCA9780.0500
Supplier: PanReac AppliChem
TRITON X100 REPLACEMENT TERGITOL 15S9 1 * 500 mL
Catalog # APPCA9780.0500
Supplier: PanReac AppliChem
Frequently Bought Together
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
Screw caps, PP, black with PE foam insert
Supplier: Kautex
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
CLASSIQSwabs™ Sterile Dry Fiber Wrapped Swabs
Supplier: Copan
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) 0.5 M in aqueous solution, autoclaved pH 8.0
Supplier: Thermo Fisher Scientific