HSD17B12 RECOMBINANT PROTEIN 1 * 50 µG
Catalog # AVIVOPCD03990-50UG
Supplier: Aviva Systems Biology
Frequently Bought Together
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
VWR®, Cryo coder cap inserts
Supplier: VWR Collection
VWR®, Cryovials
Supplier: VWR Collection
Cell culture plates, 384-well, CELLSTAR®
Supplier: Greiner Bio-One
Film for chemiluminescent imaging, Kodak BioMax® Light
Supplier: CARESTREAM HEALTH
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
Screw cap trays for 2D barcoded storage tubes
Supplier: THERMO MATRIX TECHNOLOGIES
Disposal container, Biocompact, 3L 1 * 30 items
Supplier: Greiner Bio-One
Chemical bottles, wide mouth, square
Supplier: RIXIUS
AcroPrep™ 24-Well Receiver Plate
Supplier: Cytiva (Formerly Pall Lab)
Sterile containers, Falcon®
Supplier: Corning
Microcentrifuge tubes, 0,5/1,5/2,0 ml, with/without screw caps
Supplier: Thermo Fisher Scientific
Cell culture plates and Multidishes, Nunclon™ Sphera™
Supplier: Thermo Fisher Scientific
PCR plates, 96-well, Armadillo™
Supplier: Thermo Fisher Scientific
Spare Lid of Ice Pan 9L, colour option, RED, BLUE, GREEN, BLACK 1 * 1 items
Supplier: VWR Collection