2-AMINO-5,7-DIMETHYL-1,8-NAPHTHYRIDINE 1 * 1 g
Catalog # APOSOR4484-1G
Supplier: Apollo Scientific
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
1-Butanol ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethylene glycol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Hexane (mixture of isomers), TECHNICAL
Supplier: VWR Chemicals
n-Hexane ≥99%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Glycerine 87% 86%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Pyridine ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Trifluoroacetic acid, Sigma-Aldrich®
Supplier: Merck