Phenazine-1,6-diol
Bottle, 250 mg
Catalog # APOSOR62191-250MG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
- CAS Number:69-48-7
- Formula:C₁₂H₈N₂O₂
- MDL Number:MFCD01708477
- Molecular Weight:212.20 g/mol
Specifications
Specifications
- Pack type:Bottle
- Pk:250 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



