Order Entry
Italy
Orders LinkContactUsLinkComponent
2-Hydroxy-3-methyl-5-nitropyridine 98%

2-Hydroxy-3-methyl-5-nitropyridine 98%

Bottle
Catalog # APOSOR345050-5G
Supplier:   Apollo Scientific
2-Hydroxy-3-methyl-5-nitropyridine 98%
Catalog # APOSOR345050-5G
Supplier:  Apollo Scientific
CAS Number:  21901-34-8
2-Hydroxy-3-methyl-5-nitropyridine 98%

Bottle

CAS Number:  21901-34-8
Price
24,60 €
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  3-Methyl-5-nitropyridin-2-ol, 3-Methyl-5-nitropyridin-2(1H)-one, 3-Methyl-5-nitro-2-pyridone, 2-Hydroxy-3-methyl-5-nitropyridine
  • CAS Number:21901-34-8
  • Formula:C₆H₆N₂O₃
  • MDL Number:MFCD03095073
  • Molecular Weight:154,12344 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications