Order Entry
Italy
Orders LinkContactUsLinkComponent
1,1-Dimethylurea

1,1-Dimethylurea

Bottle, 25 g
Catalog # APOSOR48247-25G
Irritant
Supplier:   Apollo Scientific
1,1-Dimethylurea
Catalog # APOSOR48247-25G
Supplier:  Apollo Scientific
CAS Number:  598-94-7
1,1-Dimethylurea

Bottle, 25 g

CAS Number:  598-94-7
Price
36,20 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  N,N-Dimethylurea
  • CAS Number:598-94-7
  • Formula:C₃H₈N₂O
  • MDL Number:MFCD00007959
  • Molecular Weight:88.11 g/mol
  • Melting Pt:178…183 °C
  • Density:1.255 g/cm³ (25 °C)
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications