PLATINUM ON ACTIVATED CARBON, 5% LOADING 1 * 25 g
Catalog # APOSOR935454-25G
Supplier: Apollo Scientific
PLATINUM ON ACTIVATED CARBON, 5% LOADING 1 * 25 g
Catalog # APOSOR935454-25G
Supplier: Apollo Scientific
Frequently Bought Together
Trifluoroacetic acid ≥99.9% for peptide synthesis
Supplier: VWR Chemicals
Ethyl acetate anhydrous, 99.8 % 1 * 2 L
Supplier: Merck
Chloroform ≥99.9% (GC, on anhydrous basis) stabilised, PESTINORM® Supra Trace
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
N,N-Dimethylacetamide anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99%
Supplier: Thermo Fisher Scientific
n-Pentane ≥99% for HPLC
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% for LC-MS
Supplier: Thermo Fisher Scientific
Acetone ≥99.5%, Laboratory Reagent
Supplier: Honeywell Chemicals