Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
STANDARD CALIBRAZIONE 0.760AW 50PZ 1 * 50 pezzi
Catalog # MSPPSQPD081792
Supplier:
MARKET SOURCE PART PROCESS
Expand image
STANDARD CALIBRAZIONE 0.760AW 50PZ 1 * 50 pezzi
Catalog # MSPPSQPD081792
Supplier:
MARKET SOURCE PART PROCESS
Frequently Bought Together
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions, ready to use, referenced at 25 °C, Certipur®
Supplier: Merck
Nitrite standard solution, 1000 mg/L NO₂- in Water (from NaNO₂) Certipur®, Supelco®
Supplier: Merck
ICP multi-element standards, CertiPUR®, Supelco®
Supplier: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck