Order Entry
Italy
Orders LinkContactUsLinkComponent
 

GC septa

BTO premium high-temperature GC septa, 10 mm, 400 °C
  SGEA041918
GC septa
  SGEA041918
GC septa
GC septa

BTO premium high-temperature GC septa, 10 mm, 400 °C

 
64,20 €
   
1 * 25 pezzi
 
 
  • Ø:
    10 mm
  • Max. temp.:
    400 °C
  • Description:
    BTO premium high-temperature GC septa
  • Seq:
    08
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.