VeriFluor™ Far-Red Passive Reference Dye
VeriFluor™ far-red passive reference dye, 400× in water, 640/660 nm, 500 µl
Catalog # 29087-500UL
Supplier: Biotium
Jump to:
Product Details & Documents
A far-red passive reference dye for qPCR that frees up the ROX channel for use with JUN™ probes, EvaRuby™ qPCR dye, and other orange dyes.
- An alternative to ROX for qPCR normalization in the red channel
- Drop-in replacement for Mustang Purple™ on most instruments
- Tolerates DTT in qPCR reactions, unlike Mustang Purple™
- More soluble than ROX for easier mixing and more accurate results
- Stand-alone dye solution for use with our Forget-Me-Not™ Master Mixes or other mixes
VeriFluor™ Far-Red is a far-red passive reference dye that is used to normalize the fluorescent reporter signal during qPCR and eliminate well-to-well variation. It is an alternative to normalization with ROX passive reference dye and a direct replacement for Mustang Purple™.
Specifications
Specifications
- Pk:500 µl
- Description:VeriFluor™ far-red passive reference dye, 400× in water
- Ex/Em:640/660 nm
- Cat. No.:29087-500UL
- Storage conditions:Store at –10...–35 °C