Order Entry
Italy
Orders LinkContactUsLinkComponent
TBX agar

TBX agar

TBX agar, 90 mm ready-to-use plates, 100 ml, 6 Bottle
Catalog # LIOF481170
Supplier:   LIOFILCHEM
TBX agar
Catalog # LIOF481170
Supplier:  LIOFILCHEM
CAS Number:  
TBX agar

TBX agar, 90 mm ready-to-use plates, 100 ml, 6 Bottle

CAS Number:  
Price
63,30 €
Log in for availability
1 * 6 Bott.
Product Details & Documents

X-glucuronide, contained in the TBX agar formulation, is the chromogenic agent that allows the determination of the β-Dglucuronidase activity, which is a highly specific enzyme for E. coli. Gram-positive bacteria are inhibited by bile salts.

Specifications

Specifications

  • Pk:
    6 Bottle
  • Description:
    TBX agar, 90 mm ready-to-use plates
  • Filling:
    100 ml

Specifications