Erlenmeyer Flasks with Screw Neck
100 ml, Amber, GL 25
Catalog # LENZ81004049
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, amber coloured, with DIN thread.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to rapid temperature changes
With white volume graduations and white marking field. Amber coloured to protect light-sensitive substances.
Specifications
- Thread:GL 25
- Capacity:100 ml
- Colour:Amber
- Seq:3