Order Entry
Italy
ContactUsLinkComponent
Dulbecco’s MEM (DMEM) High Glucose, w/ L-Glutamine, w/o Thiamine (Powder) 1 * 1 items
  USBID9803-02-10L
Dulbecco’s MEM (DMEM) High Glucose, w/ L-Glutamine, w/o Thiamine (Powder) 1 * 1 items
  USBID9803-02-10L
Frequently Bought Together