Order Entry
Italy
ContactUsLinkComponent
Meßkolben 40ml NS14 /PE ,Kl.A/KB,BRAUN Hals D. 17mm Braunglas Toleranz +-0,1ml. Kl. A/konformitätsbescheinigt 1 * 1 ST
Catalog # NEUB02935000401417
Meßkolben 40ml NS14 /PE ,Kl.A/KB,BRAUN Hals D. 17mm Braunglas Toleranz +-0,1ml. Kl. A/konformitätsbescheinigt 1 * 1 ST
Catalog # NEUB02935000401417
Frequently Bought Together