Order Entry
Italy
ContactUsLinkComponent
Objective HI PLAN I 40x/0.50 Free working distance: 2.0 mm For Cover glass thickness : 1.1mm 1 * 1 items
Catalog # LEIM11506265
Supplier:  LEICA MICROSYSTEMS
Objective HI PLAN I 40x/0.50 Free working distance: 2.0 mm For Cover glass thickness : 1.1mm 1 * 1 items
Catalog # LEIM11506265
Supplier:  LEICA MICROSYSTEMS
Frequently Bought Together