Order Entry
Italy
ContactUsLinkComponent
Objective Plan Pol 4X/0.10 NA Free working distance: 26.2mm For use with and without Cover Glass 1 * 1 items
Catalog # LEIM13613640
Supplier:  LEICA MICROSYSTEMS
Objective Plan Pol 4X/0.10 NA Free working distance: 26.2mm For use with and without Cover Glass 1 * 1 items
Catalog # LEIM13613640
Supplier:  LEICA MICROSYSTEMS
Frequently Bought Together