Order Entry
Italy
Orders LinkContactUsLinkComponent
Bottle Holders
Catalog # TEKMHWBH-0500-3
Supplier:   TEKNOMEK
Bottle Holders
Catalog # TEKMHWBH-0500-3
Supplier:  TEKNOMEK
Bottle Holders
Bottle Holders
Price
Inquire For Price
Log in for availability
1 * 1 pezzi
Product Details & Documents
Our bottle holders come in a range of sizes.
  • Brushed dull polish finish
  • Easy clean design
  • 80 mm diameter hole
Our bottle holders are the ideal solution to your storage needs, coming in a range of sizes (1, 2 or 3 bottle capacities) to suit your requirements.
The bottle holders are manufactured in fully welded 304 grade stainless steel with a brushed dull polish finish.
The stainless steel bottle holder is suitable for housing 500 ml bottles with 80 mm diameter rings, ensuring they can hold a wide variety of bottles.