Methanol ≥99.6%, Laboratory Reagent
Glass bottle, 4 L
Nome del fornitore: Honeywell Chemicals
Methanol ≥99.6%, Laboratory Reagent
Codice catalogo HONC179957-4L
Nome del fornitore: Honeywell Chemicals
Numero CAS: 67-56-1
Jump to:
Product Details & Documents
- ≥95% for general laboratory use.
Beilstein Number: 1098229
Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
- CAS Number:67-56-1
- Formula:CH₃OH
- MDL Number:MFCD00004595
- Molecular Weight:32.04 g/mol
- Merck Index:13,05984
- Boiling Pt:64…65 °C (1.013 hPa)
- Melting Pt:–98 °C
- Density:0.79 g/cm³ (20 °C)
- Flash Pt:11 °C
- Storage Temperature:Ambient
- EINECS:200-659-6
- UN:1230
- ADR:3,II
- REACH:01-2119433307-44
Documents
sds_179957
SDS
Specifications
Specification Test Results
- Explosion Limit:36 %
- Identity (IR):complying
- Assay (GC):Min. 99.6 %
- Non-volatile matter:Max. 0.01 %
- Water (Karl Fischer):Max. 0.1 %
- Free acid (as HCOOH):Max. 0.03 %
- Free alkali (as NH₃):Max. 0.01 %
- Vapor Pressure:410 mmHg ( 50 °C)
- Autoignition Temperature:725 °F
- Vapor Density:1.11 (vs air)
Specifications
- Pack type:Glass bottle
- Pk:4 L
- Cat. No.:HONC179957-4L
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health



