Chainmail Gloves
S, Silver
Catalog # PRTWAC01SIRS
Supplier: Portwest
Jump to:
Product Details & Documents
Perfect for meat processing environments when protection against sharp blades is vital.
- Made from stainless steel chainmail for maximum protection
- Ideal for meat processing
- Ambidextrous design suitable for left- or right-handed users
- Suited for food processing plants and resistant to greases, animal fats, and oils
- Can be used in food industries (metal detectable)
- Sold in singles
Clip fastening for improved hygiene to reduce bacteria build up. Ambidextrous to suit right and left handed users. Sold singly.
Colour: Silver
: EN 1082-1. CE Food Safe. CE certified.
Documents
TDS Chainmail Gloves
Datasheet
Specifications
- Glove size:S
- Colour:Silver
- Seq:01
- Cat. No.:PRTWAC01SIRS
- Latex / latex-free:Latex-free
- Disposable / reusable:Reusable
- Material:Stainless steel
- Powdered / powder-free:Powder-free
- Sterility:Non sterile
- Ambidextrous / hand-specific:Ambidextrous



