Order Entry
Italy
ContactUsLinkComponent
Cell DIVE Imager - fully-automated tissue imaging system - Five channel solid state illumination system (390/22, 470/24, 542/33, 631/28, 730/40 nm) - Penta-band polychroic beam splitter (Excitation bands: 390/23, 470/26, 542/32, 631/28, 730/39 Emis 1 * 1 items
Catalog # LEIM17429159
Supplier:  LEICA MICROSYSTEMS
Cell DIVE Imager - fully-automated tissue imaging system - Five channel solid state illumination system (390/22, 470/24, 542/33, 631/28, 730/40 nm) - Penta-band polychroic beam splitter (Excitation bands: 390/23, 470/26, 542/32, 631/28, 730/39 Emis 1 * 1 items
Catalog # LEIM17429159
Supplier:  LEICA MICROSYSTEMS
Frequently Bought Together