SURESTART cert 96 well plate, 5.6mm rnd, total v, 100ul, pp, 1 * 50 items
Catalog # HYPE60180-P210
Supplier: Thermo Fisher Scientific
SURESTART cert 96 well plate, 5.6mm rnd, total v, 100ul, pp, 1 * 50 items
Catalog # HYPE60180-P210
Supplier: Thermo Fisher Scientific
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals