Order Entry
Italy
ContactUsLinkComponent
SURESTART cert 384 plate, square, u base, 58ul, pp 1 * 60 items
Catalog # HYPE60180-P203
SURESTART cert 384 plate, square, u base, 58ul, pp 1 * 60 items
Catalog # HYPE60180-P203
Frequently Bought Together