Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SURESTART webseal mat, clear, silicone, for 96 round well plates, 8mm diameter, flat base, large penetration area, pre-slit, 1 * 5 items
Catalog # HYPE60180-M183
Supplier:
Thermo Fisher Scientific
Expand image
SURESTART webseal mat, clear, silicone, for 96 round well plates, 8mm diameter, flat base, large penetration area, pre-slit, 1 * 5 items
Catalog # HYPE60180-M183
Supplier:
Thermo Fisher Scientific
Frequently Bought Together
Avantor® ACE® C8, HPLC Columns, 5 µm
Supplier: Avantor
VWR® Hichrom Precolumn Filters
Supplier: Avantor
Level indicators, b.safe
Supplier: Bohlender
HPLC columns, NUCLEOSIL® C18
Supplier: MACHEREY-NAGEL
Accessories for optima® 4X00
Supplier: PerkinElmer