ARABINOSE AR 1VL 1 * 1 items
Catalog # HIMEDD001-1VL
Supplier: HIMEDIA
Frequently Bought Together
Cyclohexane, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Hexane ≥99% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
(±)-2-Ethyl-1-hexanol 99%
Supplier: Thermo Fisher Scientific
1,2-Dichloroethane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Acetone ≥99.5%, Laboratory Reagent
Supplier: Honeywell Chemicals
Trimethylsulphonium hydroxide 0.25 M in methanol, Supelco®
Supplier: Merck
tert-Butanol ≥99%
Supplier: Thermo Fisher Scientific
4-Methyl-2-pentanone 99+% for HPLC
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.8% (by GC), CHROMASOLV™ for liquid chromatography, Riedel-de Haën™
Supplier: Honeywell Chemicals