Reagecon BY2 Colour Reference Solution according to European Pharmacopoeia (EP) 1 * 100 mL
Catalog # REGEEPBY102
Supplier: REAGECON
Reagecon BY2 Colour Reference Solution according to European Pharmacopoeia (EP) 1 * 100 mL
Catalog # REGEEPBY102
Supplier: REAGECON
Frequently Bought Together
Starch 1% in aqueous solution stabilised Reag. Ph. Eur. 1085103
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Tin(II) chloride 30% in hydrochloric acid 20% Reag. Ph. Eur. 1085001
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH Standard Solutions, IUPAC, Certified
Supplier: Hach