Rabbit monoclonal [EPR23651-4] to IRS1 - BSA and Azide free (Capture) 1 * 100 µG
Catalog # ABCAAB280019-100
Supplier: Abcam
Rabbit monoclonal [EPR23651-4] to IRS1 - BSA and Azide free (Capture) 1 * 100 µG
Catalog # ABCAAB280019-100
Supplier: Abcam
Frequently Bought Together
VWR®, Cryoboxes, Half Size, Cardboard
Supplier: VWR Collection
Falcon® Petri Dishes, 50×9 mm, Tight-Fit Lid, Sterile, Corning
Supplier: Corning
Microplates, 96-well
Supplier: Greiner Bio-One
Cryoboxes, 133×133 mm
Supplier: TENAK
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
Transfer pipettes
Supplier: RATIOLAB
96-Well Polypropylene Storage Blocks, Corning®
Supplier: Corning
Culture media bottles, square, sterile, Nalgene®
Supplier: Thermo Fisher Scientific
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
PCR plates, 96-well, non skirted, low profile, Thermo-Fast®
Supplier: Thermo Fisher Scientific
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Blender Bags
Supplier: Grade Products
Container, conical, with screw cap, Corning® Gosselin™
Supplier: Corning
16S Barcoding Kits
Supplier: Oxford Nanopore Technologies
Storage tubes, 2D barcoded, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES