VWR®, Septa ND11
High performance GC injection port, red, with centre guide, 3,0 mm
Supplier: VWR Collection
Jump to:
Product Details & Documents
Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
- Fits for a broad range of caps
- Easy exchange
- Different hardness and septa thickness available
Septa choice depends on the application.
Specifications
- Septum:High performance GC injection port, red, with centre guide
- Seq:12
- Thickness:3,0 mm