Order Entry
Italy
Orders LinkContactUsLinkComponent
VWR®, Septa ND11
VWR®, Septa ND11
Universal HT injection port, transparent, 3,0 mm
Codice catalogo 548-0184A
Nome del fornitore:   VWR Collection
VWR®, Septa ND11
Codice catalogo 548-0184A
Nome del fornitore:  VWR Collection
VWR®, Septa ND11
VWR®, Septa ND11
Universal HT injection port, transparent, 3,0 mm
Price
40,80 €
Log in for availability
1 * 25 pezzi
Product Details & Documents
Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
  • Fits for a broad range of caps
  • Easy exchange
  • Different hardness and septa thickness available
Septa choice depends on the application.
Specifications
  • Septum:
    Universal HT injection port, transparent
  • Seq:
    11
  • Thickness:
    3,0 mm