Lithium Standard, 7000 ppm flame photometry 1 * 500 mL
Catalog # REGEFILI7
Supplier: REAGECON
Frequently Bought Together
VWR® AVS TITRINORM, Silicon Standard Solution, 1000 mg/L Si in Water, Standard for AAS
Supplier: VWR Chemicals
VWR® ARISTAR®, Arsenic Standard Solutions for ICP-MS
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Viscosity standards
Supplier: VWR Chemicals
ICP multi-element standards, CertiPUR®, Supelco®
Supplier: Merck
Buffers, concentrated solutions, Titrisol®, Supelco®
Supplier: Merck
Sumatriptan Related Compounds, Reference Standard
Supplier: USP
ICP METAL STANDARD 31 COMPONENT 1 * 100 mL
Supplier: CPACHEM
Phthalic anhydride
Supplier: EHRENSTORFER
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
ARSENIC - AS @ 1000 µG/G IN HYDROCARBON OIL 1 * 1 items
Supplier: LGC Standards PROMOCHEM
Bretylium Tosylate, Reference Standard
Supplier: USP