High Affinity Ni Charged Resin FF Prepacked Column 5 ml x 1 1 * 5 mL
Catalog # GENSL00683-51
Supplier: Genscript
High Affinity Ni Charged Resin FF Prepacked Column 5 ml x 1 1 * 5 mL
Catalog # GENSL00683-51
Supplier: Genscript
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Serological pipettes, Stripette™
Supplier: Corning
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
VWR®, Sampling Spoons
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
Supplier: VWR Collection