PROTEIN A RESIN FF PREPACK COLUMN 5ML X1 1 * 5 mL
Catalog # GENSL00680-51
Supplier: Genscript
Frequently Bought Together
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
Cell culture flasks, Falcon®
Supplier: Corning
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Supplier: EPPENDORF
Aspirating pipettes, Falcon®
Supplier: Corning
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
VWR®, Confocal Dishes for Microscopy
Supplier: VWR Collection
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Individually Wrapped Serological Pipettes, Falcon®
Supplier: Corning
VWR®, Breatheable Rayon Film Seals for Biological Cultures
Supplier: VWR Collection
96-Well clear polystyrene microplates, Corning®
Supplier: Corning
VWR®, Centrifuge Tubes, Metal-Free, Sterile
Supplier: VWR Collection
Cryoboxes, coloured, Nalgene®
Supplier: Thermo Fisher Scientific
VWR®, Transfer Pipettes, Graduated
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection