FREEZIN1 1 * 50 mL
Catalog # HIMETCL098-50ML
Supplier: HIMEDIA
Frequently Bought Together
Cryoboxes, Nalgene®
Supplier: Thermo Fisher Scientific
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR® Blender Bags
Supplier: VWR Collection
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Stoppers, PE
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Cell culture dishes, Falcon®
Supplier: Corning
Transport tubes, Axygen®
Supplier: Corning
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
n-Pentane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
U96 DeepWell™ plates 1,3 and 2,0 ml, Nunc®
Supplier: Thermo Fisher Scientific
Cell culture plates, Nunc™ Multidish
Supplier: Thermo Fisher Scientific
Collagen I-coated microplates, Corning® BioCoat®
Supplier: Corning