Order Entry
Italy
ContactUsLinkComponent
Moticam X5 Plus Attachable digital C-mount camera Sensor: CMOS Sensor size: 1/3 Live resolution: 2.0MP (1920x1080 pixels) Still resolution: 4.0MP Output: Wi-Fi (2.4 or 5 GHz), Ethernet and USB for external devices Focusable lens: 12mm 2 variable d 1 * 1 items
Catalog # MOTI1100600101322
Supplier:  MOTIC
Moticam X5 Plus Attachable digital C-mount camera Sensor: CMOS Sensor size: 1/3 Live resolution: 2.0MP (1920x1080 pixels) Still resolution: 4.0MP Output: Wi-Fi (2.4 or 5 GHz), Ethernet and USB for external devices Focusable lens: 12mm 2 variable d 1 * 1 items
Catalog # MOTI1100600101322
Supplier:  MOTIC
Frequently Bought Together