Human GABPB2 full-length ORF ( NP_005245.2, 1 a.a. - 395 a.a.) recombinant protein with GST-tag at N-terminal. 1 * 2 mg
Catalog # ABNOH00002553P01-2
Supplier: Abnova
Human GABPB2 full-length ORF ( NP_005245.2, 1 a.a. - 395 a.a.) recombinant protein with GST-tag at N-terminal. 1 * 2 mg
Catalog # ABNOH00002553P01-2
Supplier: Abnova
Frequently Bought Together
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
Cryoboxes, 133×133 mm
Supplier: TENAK
Cryogenic vials, Cryovial®
Supplier: Simport Scientific
Corning® Permeable Supports, Polycarbonate (PC) Membrane, Corning
Supplier: Corning
Serological pipettes, Stripette™
Supplier: Corning
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Cap Sealing Mats for 96 Deep Well Plates
Supplier: VWR Collection
Test tubes with screw caps
Supplier: BOETTGER PAUL
Strips of 8 PCR tubes with attached caps, 0,2 ml, Amplitube™
Supplier: Simport Scientific
Microcentrifuge tubes, 0,6/1,5/1,7/2,0/5,0 ml, SnapLock, Axygen®
Supplier: Corning
Blotting membranes, Amersham™ Hybond™-N+
Supplier: Cytiva
Poly-L-lysine-coated plates, Corning® BioCoat®, Corning
Supplier: Corning