Order Entry
Italy
ContactUsLinkComponent
Masterflex® Mechanically Coupled Centrifugal Pump Flange Kit for 1" and 1 1/4" Connections
Masterflex® Mechanically Coupled Centrifugal Pump Flange Kit for 1" and 1 1/4" Connections
Catalog # MFLX70725-70
undefined
Masterflex® Mechanically Coupled Centrifugal Pump Flange Kit for 1" and 1 1/4" Connections
Catalog # MFLX70725-70
About this item
Enhance the life of your pump with service flange kits.
  • Constructed of stainless steel (SS) parts
  • Fits 1" and 1 1/4" connections
Flange kits are competible with your 304 SS mechanically coupled centrifugal pumps.
Delivery: Includes stainless steel 150# threaded RF companion flanges, gaskets, nuts, bolts, flat washer, and lock washers.
  • Description:
    Flange Kit for 1" and 1-1/4" Connections
  • Environmentally Preferable:
  • Wetted materials:
    SS flanges, gaskets, nuts, bolts, flat washer, lockwashers
  • Warranty:
    1 year
  • Pk:
    1
Frequently Bought Together