Order Entry
Italy
ContactUsLinkComponent
Objective N PLAN L 20x/0.35 Free working distance: 6.9 mm For use with cover glass of 0 - 2 mm thickness 1 * 1 items
Codice catalogo LEIM11506247
Supplier:  LEICA MICROSYSTEMS
Objective N PLAN L 20x/0.35 Free working distance: 6.9 mm For use with cover glass of 0 - 2 mm thickness 1 * 1 items
Codice catalogo LEIM11506247
Supplier:  LEICA MICROSYSTEMS
Frequently Bought Together