Masterflex® L/S® Variable-Speed Digital Drive System with PTFE-Diaphragm Head; 90 to 260 VAC
Catalog # MFLX77915-10-IT
Supplier: Avantor Fluid Handling
CE Compliant
ISO9001
About this item
Ideal for chemical feed/metering and applications requiring high-purity, high-pressure or both.
- Flow range 80 to 800 ml/min—flow rate depends on drive rpm and tubing size
- PTFE wetted parts help maintain fluid purity
- Pump through filters or into pressurized system
- Check valve on inlet side of pump holds prime for up to two hours when pump is off
- Unique geometry minimizes fluid churning and dead volume for high metering accuracy without reagent contamination
- Accurate and repeatable settings with 3-digit LED display of rpm
- Maximize processes in a minimal footprint by stacking splash-resistant, IP33-rated drives
- Two ways to control speed, direction, and start/stop—use either the membrane keypad or remote-control capabilities
These L/S® precision variable-speed console drives offer precise ±0.25% speed control for accurate flow regulation. The large LED display and LED indicators confirm motor rpm and motor direction at a glance. The complete pump systems include all you need to start pumping.
Delivery: Includes pTFE-diaphragm pump head 07090-42, 12 ft (3.6-m) of PTFE tubing (1/4" ID x 3/8" OD), L/S drive 07528-10, two PFA pipe adapters (1/4" NPT(F) x 3/8" OD), foot valve, mounting tools, and coupling hardware.
- Maximum Flow Rate:800 ml/min
- Power:90 - 260 VAC; 50/60 Hz; 1,8 A @ 115 V; 1,2 A @ 230 V
- Accuracy:±0,25% (1 rpm at 600 and 300 rpm; 0,1 rpm at 100 rpm)
- Control type:Variable-Speed Digital
- Min. flow rate:80 ml/min
- No. of channels:1
- Pump series:L/S
- Description:L/S PTFE-Diaphragm Pump System; 6 to 600 rpm, 90 to 260 VAC
- Height:21,59 cm
- Length:43,94 cm
- Environmentally Preferable:
- Duty cycle:Continuous
- Drive IP rating:IP 33
- Min. RPM:6 rpm
- Open-head sensor:No
- Remote I/O:4 to 20 mA, 0 to 10 V, start/stop and direction via contact closure
- Tubing sizes accepted:L/S® 13, 14, 16, 25, 17, 18
- Washdown capable:No
- Tubing:06605-15, 12-ft (PTFE Tubing, 1/4" x 3/8")
- Drive:07528-10 (L/S® Variable-Speed Digital Drive with Remote I/O, 600 rpm)
- Warranty:2 years
- Pk:1
- Pump head:07090-42 (L/S® PTFE-Diaphragm Pump Head)
- Width:21,08 cm
- Housing material:ABS plastic and aluminium
- Electrical plug type:IT
- Max. RPM:600 rpm
Frequently Bought Together