METHYLAMINE (CA. 7% IN N,N-DIMETHYLFORM 1 * 500 mL
TCIAM3340-500ML
: TCI
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Dichloromethane stabilised Ph. Eur.
: VWR Chemicals
Toluene ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
: MERCK PRODUCTION CHEMICALS
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
N,N-Dimethylformamide ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Hexane (mixture of isomers), HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Petroleum spirit, 30…40 °C, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
: MACRON AVANTOR BRAND